siRNA Details
Gene Symbol: Gdpd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201352.2 | NP_958740.2 | 10 | 1510 | ||
GenBank mRNA | AK141189.1 | - | 10 | 1543 | ||
AK154771.1 | - | 10 | 1647 | |||
BC024955.1 | - | 9 | 796 | 4953174 | ||
BC026428.1 | - | 6 | 443 | 4224574 | ||
BC082585.1 | - | 10 | 1510 | 30608279 | ||
BC119442.1 | - | 1 | 169 | |||
BC119443.1 | - | 1 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
