siRNA Details
Gene Symbol: P2ry6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183168.2 | NP_898991.1 | 3 | 622 | ||
GenBank mRNA | AK142680.1 | - | 3 | 647 | ||
AX775901.1 | - | 3 | 582 | |||
BC027331.1 | - | 3 | 618 | 3982042 | ||
BC064095.1 | - | 3 | 602 | 1054566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
