siRNA Details
Gene Symbol: Plekha7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040271.1 | - | 24 | 3552 | ||
AK146866.1 | - | 3743 | ||||
BC060678.1 | - | 23 | 3473 | |||
EU380771.1 | - | 3769 | ||||
EU380772.1 | - | 3827 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
