siRNA Details
Gene Symbol: BC030336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164580.1 | NP_001158052.1 | 2 | 428 | ||
GenBank mRNA | AK052350.1 | - | 2 | 279 | ||
AK162324.1 | - | 1 | 121 | |||
BC030336.1 | - | 2 | 312 | 5401060 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006230181.
Gene Ontology Categories:
