siRNA Details
Gene Symbol: BC030336 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164580.1 | NP_001158052.1 | 2 | 425 | ||
GenBank mRNA | AK052350.1 | - | 2 | 276 | ||
AK162324.1 | - | 1 | 118 | |||
BC030336.1 | - | 2 | 309 | 5401060 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
