siRNA Details
Gene Symbol: Cog7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033318.3 | NP_001028490.2 | 10 | 1477 | ||
GenBank mRNA | AK030709.1 | - | 10 | 1444 | ||
AK145163.1 | - | 10 | 1475 | |||
AK154693.1 | - | 10 | 1479 | |||
AK163143.1 | - | 1497 | ||||
AK172200.1 | - | 10 | 1479 |
