siRNA Details
Gene Symbol: Ino80e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153580.1 | NP_705808.1 | 1 | 179 | ||
GenBank mRNA | BC026475.1 | - | 1 | 179 | 4489168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
