siRNA Details
Gene Symbol: Kctd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172747.2 | NP_766335.1 | 6 | 971 | ||
GenBank mRNA | AF534881.3 | - | 6 | 957 | ||
AK075722.1 | - | 6 | 972 | |||
BC079607.1 | - | 6 | 964 | 30359035 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
