siRNA Details
Gene Symbol: BC017158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145590.2 | NP_663565.2 | 11, 12 | 1277 | ||
GenBank mRNA | AK148236.1 | - | 11, 12 | 1268 | ||
AK149645.1 | - | 11, 12 | 1256 | |||
AK164488.1 | - | 11, 12 | 1255 | |||
AK165287.1 | - | 9, 10 | 2720 | |||
AK170611.1 | - | 10, 11 | 1340 | |||
BC017158.1 | - | 11, 12 | 1259 | 4919869 | ||
BC024080.1 | - | 10, 11 | 887 | |||
BC061499.1 | - | 1, 2 | 1203 | |||
BC094040.1 | - | 11, 12 | 1261 |
