siRNA Details
Gene Symbol: Ppfia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033319.2 | NP_001028491.1 | 16 | 2223 | ||
NM_001195086.1 | NP_001182015.1 | 17 | 2298 | |||
GenBank mRNA | BC038349.1 | - | 6 | 616 | ||
BC157936.1 | - | 17 | 2115 | 9088116 | ||
BC158007.1 | - | 16 | 2180 | 9007315 | ||
BC172096.1 | - | 17 | 2118 | 9054382 | ||
EU568869.1 | - | 17 | 2298 |

More Information
Other Species Targeted

Gene Ontology Categories:
