siRNA Details
Gene Symbol: Zfp958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145591.4 | NP_663566.1 | 4 | 331 | ||
GenBank mRNA | AK037083.1 | - | 4 | 332 | ||
AK037596.1 | - | 4 | 334 | |||
BC003267.1 | - | 4 | 334 | 3499094 | ||
BC138223.1 | - | 4 | 278 | 8861242 | ||
BC145996.1 | - | 4 | 278 | 40131299 |
