siRNA Details
Gene Symbol: Pcid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178708.3 | NP_848823.2 | 9 | 1126 | ||
GenBank mRNA | AK042945.1 | - | 9 | 616 | ||
AK045796.1 | - | 9 | 606 | |||
AK048722.1 | - | 9 | 617 | |||
AK082891.1 | - | 9 | 1192 | |||
AK163697.1 | - | 9 | 613 | |||
BC006047.1 | - | 9 | 615 | |||
BC072621.1 | - | 6 | 1060 | |||
BC118533.1 | - | 9 | 1126 | 40095328 |

More Information
Other Species Targeted

Gene Ontology Categories:
