siRNA Details
Gene Symbol: Tti2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199988.1 | NP_001186917.1 | 5 | 1721 | ||
NM_144927.3 | NP_659176.2 | 5 | 1721 | |||
GenBank mRNA | AK045392.1 | - | 5 | 1726 | ||
AK046031.1 | - | 5 | 1711 | |||
AK084242.1 | - | 5 | 1711 | |||
AK139425.1 | - | 5 | 1698 | |||
BC019943.1 | - | 5 | 1424 | 4017819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
