siRNA Details
Gene Symbol: Fgl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145594.2 | NP_663569.2 | 5, 6 | 605 | ||
GenBank mRNA | AF478470.1 | - | 5, 6 | 615 | ||
BC021946.1 | - | 5, 6 | 573 | 5098805 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
