siRNA Details
Gene Symbol: Helt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173789.4 | NP_776150.2 | 4 | 1268 | ||
GenBank mRNA | AB098077.1 | - | 4 | 719 | ||
AB120968.1 | - | 4 | 1268 | |||
AK044070.1 | - | 3 | 819 | |||
BC103599.1 | - | 4 | 1153 | 40045925 | ||
BC103600.1 | - | 4 | 1153 | 40045926 | ||
BC103601.1 | - | 4 | 1153 | 40045923 | ||
DQ294234.1 | - | 4 | 731 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
