siRNA Details
Gene Symbol: Psd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027626.1 | NP_081902.1 | 5 | 768 | ||
NM_030263.5 | NP_084539.2 | 10 | 1110 | |||
NM_177698.4 | NP_808366.2 | 13 | 2874 | |||
GenBank mRNA | AB220685.1 | - | 13 | 2874 | ||
AK031477.1 | - | 5 | 711 | |||
AK122405.1 | - | 4 | 424 | |||
AK147606.1 | - | 5 | 768 | |||
AK161498.1 | - | 10 | 1096 | |||
BC042208.1 | - | 1 | 97 | |||
BC052365.1 | - | 10 | 1075 | |||
BC131913.1 | - | 5 | 680 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
