siRNA Details
Gene Symbol: Psd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030263.5 | NP_084539.2 | 2 | 306 | ||
NM_177698.4 | NP_808366.2 | 4 | 2067 | |||
GenBank mRNA | AB220685.1 | - | 4 | 2067 | ||
AK016466.1 | - | 2 | 361 | |||
AK028684.1 | - | 5 | 1879 | |||
AK161498.1 | - | 2 | 292 | |||
BC052365.1 | - | 2 | 270 | |||
CJ047427.1 | - | 2 | 307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
