siRNA Details
Gene Symbol: Zfp930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013379.2 | NP_001013397.2 | 4 | 588 | ||
GenBank mRNA | AK040352.1 | - | 2 | 1901 | ||
D10627.1 | - | 1 | 192 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013379.2 | NP_001013397.2 | 4 | 588 | ||
GenBank mRNA | AK040352.1 | - | 2 | 1901 | ||
D10627.1 | - | 1 | 192 |