siRNA Details
Gene Symbol: Gatad2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113346.1 | NP_001106817.1 | 5 | 800 | ||
NM_001286450.1 | NP_001273379.1 | 5 | 948 | |||
NM_145596.4 | NP_663571.2 | 5 | 948 | |||
GenBank mRNA | AK089074.1 | - | 5 | 930 | ||
AK160058.1 | - | 5 | 797 | |||
BC038221.1 | - | 5 | 698 | 5321913 | ||
CN666280.1 | - | 3 | 318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
