siRNA Details
Gene Symbol: Klhl26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122830.1 | NP_001116302.1 | 2 | 1560 | ||
NM_178771.3 | NP_848886.1 | 3 | 1743 | |||
GenBank mRNA | AK036321.1 | - | 3 | 1585 | ||
AK041089.1 | - | 3 | 1746 | |||
AK048601.1 | - | 3 | 1711 | |||
AK049946.1 | - | 3 | 1743 | |||
AK163170.1 | - | 3 | 1743 | |||
BC023909.1 | - | 3 | 1752 | 5322205 | ||
BC025484.1 | - | 3 | 1654 | 5151231 | ||
BC098185.1 | - | 3 | 1718 | 3594528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
