siRNA Details
Gene Symbol: Tmem184c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145599.4 | NP_663574.3 | 6 | 1065 | ||
GenBank mRNA | AK033404.1 | - | 6 | 1034 | ||
AK164182.1 | - | 6 | 1033 | |||
BC004056.1 | - | 6 | 1023 | 3589819 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
