siRNA Details
Gene Symbol: Rtbdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144929.2 | NP_659178.1 | 3 | 978 | ||
GenBank mRNA | AK039726.1 | - | 1 | 1485 | ||
AK080715.1 | - | 3 | 294 | |||
AY428631.1 | - | 2 | 190 | |||
AY527275.1 | - | 3 | 978 | |||
AY527276.1 | - | 4 | 433 | |||
BC022744.1 | - | 2 | 550 | 4527012 | ||
BC023051.1 | - | 2 | 506 | 5361063 | ||
BC027079.1 | - | 2 | 506 | 5357069 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
