siRNA Details
Gene Symbol: Txnl4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175646.3 | NP_783577.2 | 2, 3 | 285 | ||
GenBank mRNA | AK079533.1 | - | 2, 3 | 384 | ||
AK085226.1 | - | 2, 3 | 266 | |||
BC094379.1 | - | 2, 3 | 301 | 30541506 | ||
BC132373.1 | - | 1, 2 | 210 | 40130650 | ||
BC138333.1 | - | 1, 2 | 210 | 8861353 | ||
BY205801.1 | - | 2, 3 | 279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
