siRNA Details
Gene Symbol: Plcg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172285.1 | NP_758489.1 | 27 | 3115 | ||
GenBank mRNA | AK142173.1 | - | 27 | 3118 | ||
AK170430.1 | - | 27 | 3125 | |||
BC019654.1 | - | 14 | 1649 | |||
BC023877.1 | - | 27 | 3115 | 5354925 | ||
BC037700.1 | - | 9 | 907 | |||
BC049273.1 | - | 10 | 1041 |
