siRNA Details
Gene Symbol: Klhl36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146219.1 | NP_666331.1 | 2 | 207 | ||
GenBank mRNA | AK131135.1 | - | 2 | 132 | ||
AK157027.1 | - | 2 | 241 | |||
AK164240.1 | - | 2 | 247 | |||
AK164831.1 | - | 2 | 241 | |||
BC025816.1 | - | 2 | 207 | 5098064 |
