siRNA Details
Gene Symbol: Mthfsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166482.1 | NP_001159954.1 | 7, 8 | 762 | ||
NM_172761.3 | NP_766349.1 | 7, 8 | 856 | |||
GenBank mRNA | AK083317.1 | - | 7, 8 | 796 | ||
AK141278.1 | - | 7, 8 | 691 | |||
AK188342.1 | - | 2, 3 | 163 | |||
BC023782.1 | - | 7, 8 | 833 | |||
BC026652.1 | - | 6, 7 | 732 | |||
BC052066.1 | - | 6, 7 | 2139 | 5708660 |

More Information
Other Species Targeted

Gene Ontology Categories:
