siRNA Details
Gene Symbol: Mthfsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166482.1 | NP_001159954.1 | 4 | 398 | ||
NM_172761.3 | NP_766349.1 | 4 | 492 | |||
GenBank mRNA | AK083317.1 | - | 4 | 432 | ||
AK141278.1 | - | 4 | 327 | |||
BC023782.1 | - | 4 | 469 | |||
BC026652.1 | - | 3 | 368 | |||
BC052066.1 | - | 4 | 398 | 5708660 | ||
BQ444001.1 | - | 4 | 398 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
