siRNA Details
Gene Symbol: Klhdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145605.2 | NP_663580.2 | 10 | 1275 | ||
GenBank mRNA | AK089954.1 | - | 10 | 1239 | ||
AK161674.1 | - | 10 | 1251 | |||
BC012312.1 | - | 10 | 1275 | 2631632 | ||
BC023738.1 | - | 10 | 1246 | 5352770 | ||
BC058359.1 | - | 10 | 1223 | 6815650 |
