siRNA Details
Gene Symbol: Med17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144933.1 | NP_659182.1 | 4 | 772 | ||
GenBank mRNA | AK033151.1 | - | 4 | 778 | ||
AK038451.1 | - | 4 | 812 | |||
AK049112.1 | - | 4 | 812 | |||
AK142033.1 | - | 4 | 811 | |||
AK151377.1 | - | 4 | 812 | |||
BC004062.1 | - | 3 | 449 | |||
BC020185.1 | - | 4 | 772 | 4505959 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
