siRNA Details
Gene Symbol: Med17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144933.1 | NP_659182.1 | 8, 9 | 1404 | ||
GenBank mRNA | AK033151.1 | - | 8, 9 | 1410 | ||
AK038451.1 | - | 8, 9 | 1444 | |||
AK049112.1 | - | 8, 9 | 1444 | |||
AK142033.1 | - | 8, 9 | 1443 | |||
AK151377.1 | - | 8, 9 | 1444 | |||
BC004062.1 | - | 7, 8 | 1081 | |||
BC020185.1 | - | 8, 9 | 1404 | 4505959 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
