siRNA Details
Gene Symbol: Kank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145611.4 | NP_663586.3 | 3 | 1366 | ||
GenBank mRNA | AK031324.1 | - | 3 | 1359 | ||
AK045492.1 | - | 3 | 1338 | |||
AK049288.1 | - | 3 | 1367 | |||
BC006647.1 | - | 3 | 1354 | |||
BC010245.1 | - | 3 | 1323 | 3710765 |
