siRNA Details
Gene Symbol: Cypt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173412.2 | NP_775588.2 | 1 | 584 | ||
GenBank mRNA | AF463501.1 | - | 1 | 584 | ||
AK005791.1 | - | 1 | 577 | |||
BC048530.1 | - | 1 | 573 | |||
BC060952.1 | - | 1 | 573 | 6772242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
