siRNA Details
Gene Symbol: Nfrkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172766.3 | NP_766354.2 | 15, 16 | 1678 | ||
GenBank mRNA | AK041206.1 | - | 15, 16 | 1916 | ||
AK049804.1 | - | 1, 2 | 53 | |||
BC033595.1 | - | 15, 16 | 1668 | 4947357 | ||
BC059861.1 | - | 15, 16 | 1725 | |||
BC059900.1 | - | 6, 7 | 1325 | |||
BC060076.1 | - | 15, 16 | 1907 |
