siRNA Details
Gene Symbol: Foxred1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291448.1 | NP_001278377.1 | 4 | 626 | ||
NM_001291449.1 | NP_001278378.1 | 4 | 631 | |||
NM_172291.2 | NP_758495.1 | 4 | 626 | |||
GenBank mRNA | AK054231.1 | - | 4 | 518 | ||
AK144759.1 | - | 4 | 552 | |||
AK163722.1 | - | 4 | 485 | |||
BC017604.1 | - | 4 | 584 | |||
BC024806.1 | - | 4 | 496 | 5361053 | ||
BC027386.1 | - | 2 | 1200 | |||
BC028892.1 | - | 2 | 1597 |
