siRNA Details
Gene Symbol: Scn3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083917.1 | NP_001077386.1 | 3, 4 | 626 | ||
NM_001286614.1 | NP_001273543.1 | 3, 4 | 622 | |||
NM_178227.4 | NP_839941.1 | 3, 4 | 626 | |||
GenBank mRNA | AK049286.1 | - | 3, 4 | 624 | ||
AK049747.1 | - | 3, 4 | 445 | |||
AK076466.1 | - | 2, 3 | 270 | |||
AK173115.1 | - | 3, 4 | 503 | |||
AY049036.1 | - | 2, 3 | 231 | |||
BC032253.1 | - | 3, 4 | 420 | 5368626 | ||
BC053919.1 | - | 2, 3 | 521 | 6308278 | ||
BC058083.1 | - | 3, 4 | 565 | |||
BC058636.1 | - | 3, 4 | 625 | 5702879 | ||
CB520894.1 | - | 3, 4 | 626 |

More Information
Other Species Targeted

Gene Ontology Categories:
