siRNA Details
Gene Symbol: Gramd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172768.1 | NP_766356.1 | 15 | 2109 | ||
GenBank mRNA | AK028277.1 | - | 16 | 2004 | ||
AK033156.1 | - | 15 | 2109 | |||
AK122465.1 | - | 14 | 2069 | |||
BC030911.1 | - | 6 | 853 | |||
BC070451.1 | - | 14 | 1943 | 30616509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
