siRNA Details
Gene Symbol: Rnf214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178709.4 | NP_848824.1 | 9, 10 | 1768 | ||
GenBank mRNA | AK028768.1 | - | 9, 10 | 1403 | ||
AK030208.1 | - | 9, 10 | 1400 | |||
AK032376.1 | - | 9, 10 | 834 | |||
AK033867.1 | - | 9, 10 | 1264 | |||
AK051527.1 | - | 9, 10 | 1769 | |||
AK078706.1 | - | 4, 5 | 302 | |||
AK136229.1 | - | 4, 5 | 293 | |||
BC023073.1 | - | 1, 2 | 46 | |||
BC049909.1 | - | 9, 10 | 909 | |||
BC094247.1 | - | 8, 9 | 1253 | 5717984 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
