siRNA Details
Gene Symbol: Rnf214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178709.4 | NP_848824.1 | 9, 10 | 1777 | ||
GenBank mRNA | AK028768.1 | - | 9, 10 | 1412 | ||
AK030208.1 | - | 9, 10 | 1409 | |||
AK032376.1 | - | 9, 10 | 843 | |||
AK033867.1 | - | 9, 10 | 1273 | |||
AK051527.1 | - | 9, 10 | 1778 | |||
AK078706.1 | - | 4, 5 | 311 | |||
AK136229.1 | - | 4, 5 | 302 | |||
BC023073.1 | - | 1, 2 | 55 | |||
BC049909.1 | - | 9, 10 | 918 | |||
BC094247.1 | - | 8, 9 | 1262 | 5717984 |
