siRNA Details
Gene Symbol: Rnf214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178709.4 | NP_848824.1 | 13, 14 | 2473 | ||
GenBank mRNA | AK028768.1 | - | 13, 14 | 2108 | ||
AK030208.1 | - | 13, 14 | 2105 | |||
AK032376.1 | - | 13, 14 | 1539 | |||
AK033867.1 | - | 12, 13 | 2805 | |||
AK051527.1 | - | 13, 14 | 2474 | |||
AK078706.1 | - | 8, 9 | 1007 | |||
AK136229.1 | - | 9, 10 | 1103 | |||
BC023073.1 | - | 6, 7 | 856 | |||
BC049909.1 | - | 14, 15 | 1719 | |||
BC064100.1 | - | 3, 4 | 589 | |||
BC094247.1 | - | 12, 13 | 1958 | 5717984 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
