siRNA Details
Gene Symbol: Zbtb16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033324.2 | NP_001028496.1 | 2 | 564 | ||
GenBank mRNA | AK040211.1 | - | 2 | 557 | ||
AK142947.1 | - | 2 | 565 | |||
BC132441.1 | - | 1 | 252 | |||
BC132443.1 | - | 1 | 252 | |||
BC138775.1 | - | 1 | 375 | 8861797 | ||
Z47205.1 | - | 2 | 605 |
