siRNA Details
Gene Symbol: Zbtb16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033324.2 | NP_001028496.1 | 4 | 1652 | ||
GenBank mRNA | AK040211.1 | - | 4 | 1645 | ||
AK142947.1 | - | 4 | 1653 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zbtb16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033324.2 | NP_001028496.1 | 4 | 1652 | ||
GenBank mRNA | AK040211.1 | - | 4 | 1645 | ||
AK142947.1 | - | 4 | 1653 |