siRNA Details
Gene Symbol: Snx33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175483.5 | NP_780692.2 | 1 | 2331 | ||
GenBank mRNA | AK048566.1 | - | 1 | 836 | ||
AK053761.1 | - | 1 | 866 | |||
AK142274.1 | - | 1 | 2332 | |||
BC096472.1 | - | 1 | 1081 | 3501525 |
