siRNA Details
Gene Symbol: Lman1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199222.3 | NP_954692.2 | 13 | 1556 | ||
GenBank mRNA | AB211986.1 | - | 13 | 1477 | ||
BC020188.1 | - | 11 | 1371 | 4925160 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
