siRNA Details
Gene Symbol: Gtf2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039519.2 | NP_001034608.1 | 4 | 372 | ||
NM_001282083.1 | NP_001269012.1 | 4 | 663 | |||
GenBank mRNA | AK050621.1 | - | 4 | 641 | ||
AK164255.1 | - | 5 | 805 | |||
AK165466.1 | - | 4 | 370 | |||
AK169778.1 | - | 4 | 339 | |||
BC028551.1 | - | 5 | 493 | 1495874 | ||
BC048705.1 | - | 4 | 578 | 6505667 | ||
BG088284.2 | - | 4 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
