siRNA Details
Gene Symbol: Pxylp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289645.1 | NP_001276574.1 | 7 | 1662 | ||
NM_001289646.1 | NP_001276575.1 | 7 | 1524 | |||
NM_001289647.1 | NP_001276576.1 | 6 | 1426 | |||
NM_153420.3 | NP_700469.1 | 7 | 1644 | |||
GenBank mRNA | AK036973.1 | - | 7 | 1663 | ||
AK043758.1 | - | 6 | 1382 | |||
AK082059.1 | - | 6 | 1382 | |||
AK083712.1 | - | 6 | 1363 | |||
AK084605.1 | - | 6 | 1382 | |||
AK144699.1 | - | 7 | 1484 | |||
AK164254.1 | - | 6 | 1353 | |||
BC023960.1 | - | 7 | 1593 | 5323284 | ||
BC036342.1 | - | 6 | 1371 | 4948271 |
