siRNA Details
Gene Symbol: Dnajc13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163026.1 | NP_001156498.1 | 30 | 3563 | ||
GenBank mRNA | AB490693.1 | - | 30 | 3333 | ||
CO045605.1 | - | 4 | 404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
