siRNA Details
Gene Symbol: Glyctk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039586.1 | NP_001034675.1 | 5 | 1602 | ||
NM_174846.4 | NP_777271.3 | 4 | 1738 | |||
GenBank mRNA | AF455785.1 | - | 5 | 1566 | ||
AK049425.1 | - | 4 | 1719 | |||
AK050182.1 | - | 5 | 1592 | |||
AK052709.1 | - | 4 | 1677 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
