siRNA Details
Gene Symbol: Glyctk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039586.1 | NP_001034675.1 | 5 | 949 | ||
NM_174846.4 | NP_777271.3 | 4 | 1085 | |||
GenBank mRNA | AF455785.1 | - | 5 | 918 | ||
AK049425.1 | - | 4 | 1066 | |||
AK050182.1 | - | 5 | 939 | |||
AK052709.1 | - | 4 | 1024 | |||
BC025834.1 | - | 4 | 1054 | 5132437 | ||
BC025935.1 | - | 1046 | 5097665 | |||
BC030732.1 | - | 4 | 767 | 5102500 | ||
BC030736.1 | - | 3 | 334 | |||
BC033063.1 | - | 4 | 1061 | 5100009 | ||
BC036136.1 | - | 906 | 4971642 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
