siRNA Details
Gene Symbol: Nbeal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183276.2 | NP_899099.2 | 50 | 7851 | ||
GenBank mRNA | AK129158.1 | - | 25 | 4119 | ||
BC157956.1 | - | 50 | 7832 | 9007284 | ||
BC172118.1 | - | 49 | 7751 | 9054404 |
