siRNA Details
Gene Symbol: Prss50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146227.4 | NP_666339.2 | 1 | 393 | ||
GenBank mRNA | AF481144.1 | - | 1 | 379 | ||
AK045168.1 | - | 1 | 394 | |||
BC150686.1 | - | 1 | 392 | 9087597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
